APO455837666 Apollo Scientific Ltd

6-(Trifluoromethyl)nicotinamidoxime

  • MDL number MFCD06245538
  • Cas Number : 386704-16-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 192-195
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=5468

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=5468
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart