APO455837668 Apollo Scientific Ltd

4'-(2-Chloro-6-fluorobenzyloxy)acetophenone

  • MDL number MFCD02179827
  • Cas Number : 306934-77-0
  • Molecular Weight : 278.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 92-95
Synonym 1-[4-(2-Chloro-6-fluorobenzyloxy)phenyl]ethan-1-one
Purity 97
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-2-chloro-6-fluorobenzyloxyacetophenone

Safety Information

signalword Warning
pcodes P280;P301+P312;P305+P351+P338
MSDS https://store.apolloscientific.co.uk/product/4-2-chloro-6-fluorobenzyloxyacetophenone
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart