APO455837671 Apollo Scientific Ltd

2-Chloro-4-fluorophenylacetamidoxime

  • MDL number MFCD02180957
  • Cas Number : 306937-33-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 149-151
Synonym 2-(2-chloro-4-fluorophenyl)-N'-hydroxyethanimidamide
Purity 97
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=2721

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=2721
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart