APO455837673 Apollo Scientific Ltd

1H,1H-Nonadecafluoro-3,6,9-trioxatridecan-1-ol

  • MDL number MFCD03094406
  • Cas Number : 317817-24-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-Hydroxy-1H,1H-nonadecafluoro-3,6,9-trioxatridecane, 2,2-Difluoro-2-{1,1,2,2-tetrafluoro-2-[1,1,2,2-tetrafluoro-2-(nonafluorobutoxy)ethoxy]ethoxy}ethan-1-ol
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 175-176
Product Link http://www.apolloscientific.co.uk/display_item.php?id=6015

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=6015
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart