APO455837675 Apollo Scientific Ltd

1,2,3,5-Tetrafluorobenzene

  • MDL number MFCD00000302
  • Cas Number : 2367-82-0
  • Molecular Weight : 150.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point -48
Purity 97
Density 1.39
Flash Point 4
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 82-83
Product Link https://store.apolloscientific.co.uk/product/1235-tetrafluorobenzene

Safety Information

signalword Danger
pcodes P210;P280
MSDS https://store.apolloscientific.co.uk/product/1235-tetrafluorobenzene
pictograms GHS02;GHS07
hstatements H225;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart