APO455837677 Apollo Scientific Ltd

Methyl 2,6-dichloro-5-fluoronicotinate

  • MDL number MFCD02180634
  • Cas Number : 189281-66-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Methyl 2,6-dichloro-5-fluoropyridine-3-carboxylate, 2,6-Dichloro-3-fluoro-5-(methoxycarbonyl)pyridine
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 84-86/0.1mm
Product Link https://store.apolloscientific.co.uk/product/methyl-26-dichloro-5-fluoronicotinate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-26-dichloro-5-fluoronicotinate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart