APO455837678 Apollo Scientific Ltd

5,6,7,8-Tetrahydro-2-thio-4-(trifluoromethyl)quinoline-3-carbonitrile

  • MDL number MFCD02180672
  • Cas Number : 306935-92-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 216-220
Synonym 3-Cyano-5,6,7,8-tetrahydro-2-thio-4-(trifluoromethyl)quinoline
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=4737

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=4737
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart