APO455837680 Apollo Scientific Ltd

1-[4-(Trifluoromethyl)pyrimidin-2-yl]piperidine-4-carbonyl chloride

  • MDL number MFCD02091616
  • Cas Number : 306934-79-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 158/2mmHg
Product Link http://www.apolloscientific.co.uk/display_item.php?id=5487

Safety Information

signalword Danger
msds http://www.apolloscientific.co.uk/display_item.php?id=5487
pictograms GHS05
Availability details for
Quantity
  • Availability
  • Add to Cart