APO455837681 Apollo Scientific Ltd

2-Chloro-3-(trifluoromethyl)benzoic acid

  • MDL number MFCD00792415
  • Cas Number : 39226-97-6
  • Molecular Weight : 224.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 132-134
Synonym 3-Carboxy-2-chlorobenzotrifluoride
Purity 98
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-chloro-3-trifluoromethylbenzoic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloro-3-trifluoromethylbenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart