APO455837691 Apollo Scientific Ltd

3',4',5'-Trifluoroacetophenone

  • MDL number MFCD00083533
  • Cas Number : 220141-73-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Flash Point >100
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=4975

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=4975
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart