APO455837694 Apollo Scientific Ltd

4-(1,1,2,2-Tetrafluoroethoxy)aniline

  • MDL number MFCD00042377
  • Cas Number : 713-62-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Density 1.379
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 104-108/11mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=4805

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=4805
pictograms GHS06
hstatements H301+H311;H331
Availability details for
Quantity
  • Availability
  • Add to Cart