APO455837698 Apollo Scientific Ltd

3-Fluoro-5-nitrobenzonitrile

  • MDL number MFCD05864356
  • Cas Number : 110882-60-5
  • Molecular Weight : 166.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 49-54
Synonym 3-Cyano-5-fluoronitrobenzene
Density 1.41
Flash Point 108
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-fluoro-5-nitrobenzonitrile

Safety Information

signalword Danger
pcodes P260;P280;P312
MSDS https://store.apolloscientific.co.uk/product/3-fluoro-5-nitrobenzonitrile
pictograms GHS07;GHS06
hstatements H302+H312+H332;H301+H311+H331
Availability details for
Quantity
  • Availability
  • Add to Cart