APO455837704 Apollo Scientific Ltd

4-Fluoro-3-methoxybenzonitrile

  • MDL number MFCD01569371
  • Cas Number : 243128-37-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 108-109
Synonym 5-Cyano-2-fluoroanisole
Purity 98
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 96-98/1.5mmHg
Product Link http://www.apolloscientific.co.uk/display_item.php?id=1879

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=1879
pictograms GHS07
hstatements H302;H312;H319;H332;H335
Availability details for
Quantity
  • Availability
  • Add to Cart