APO455837712 Apollo Scientific Ltd

5-Amino-1-(4-fluorophenyl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD01569434
  • Cas Number : 187949-90-2
  • Molecular Weight : 221.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Amino-4-carboxy-1-(4-fluorophenyl)-1H-pyrazole, 1-(5-Amino-4-carboxy-1H-pyrazol-1-yl)-4-fluorobenzene
Flash Point >110
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-amino-1-4-fluorophenyl-1h-pyrazole-4-carboxylic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-amino-1-4-fluorophenyl-1h-pyrazole-4-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart