APO455837713 Apollo Scientific Ltd

Tetrafluoroisophthalonitrile

  • MDL number MFCD00013290
  • Cas Number : 2377-81-3
  • Molecular Weight : 200.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 76-79
Iupac Name 2,4,5,6-tetrafluoroisophthalonitrile
Inchi Code 1S/C8F4N2/c9-5-3(1-13)6(10)8(12)7(11)4(5)2-14
Inchi Key WVHMPQKZPHOCRD-UHFFFAOYSA-N

Description

Synonym Perfluoroisophthalonitrile, 2,4,5,6-Tetrafluorobenzene-1,3-dicarbonitrile
Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 76-79
Product Link https://store.apolloscientific.co.uk/product/tetrafluoroisophthalonitrile

Safety Information

signalword Danger
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/tetrafluoroisophthalonitrile
pictograms GHS06
hstatements H301;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart