APO455837714 Apollo Scientific Ltd

4-Amino-5-(4-fluorophenyl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD00269268
  • Cas Number : 61019-25-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=2307

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=2307
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart