APO455837721 Apollo Scientific Ltd

2-Bromo-4-fluorophenacyl bromide

  • MDL number MFCD11847026
  • Cas Number : 594810-90-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2,2'-Dibromo-4-fluoroacetophenone, 2-Bromo-1-(2-bromo-4-fluorophenyl)ethan-1-one
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=47014

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=47014
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart