APO455837734 Apollo Scientific Ltd

1-(4-Fluoro-2-nitrophenyl)piperazine

  • MDL number MFCD00798371
  • Cas Number : 243128-46-3
  • Molecular Weight : 225.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Fluoro-2-(piperazin-1-yl)nitrobenzene
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-4-fluoro-2-nitrophenylpiperazine

Safety Information

signalword Warning
pcodes P280;P301+P312;P304+P340
MSDS https://store.apolloscientific.co.uk/product/1-4-fluoro-2-nitrophenylpiperazine
pictograms GHS07
hstatements H302+H312+H332;H315;H319
Availability details for
Quantity
  • Availability
  • Add to Cart