APO455837735 Apollo Scientific Ltd

2-Bromo-3-fluorobenzyl bromide

  • MDL number MFCD12911576
  • Cas Number : 1184918-22-6
  • Molecular Weight : 267.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Bromo-1-(bromomethyl)-3-fluorobenzene, alpha,2-Dibromo-3-fluorotoluene
Purity 99
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-3-fluorobenzyl-bromide

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-3-fluorobenzyl-bromide
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart