APO455837740 Apollo Scientific Ltd

Tetrafluorosuccinyl chloride

  • MDL number MFCD00013653
  • Cas Number : 356-15-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 85-87
Product Link http://www.apolloscientific.co.uk/display_item.php?id=4876

Safety Information

signalword Danger
msds http://www.apolloscientific.co.uk/display_item.php?id=4876
pictograms GHS05
Availability details for
Quantity
  • Availability
  • Add to Cart