APO455837747 Apollo Scientific Ltd

3-Chloro-5-(trifluoromethyl)benzonitrile

  • MDL number MFCD04115854
  • Cas Number : 693245-52-2
  • Molecular Weight : 205.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Chloro-5-cyanobenzotrifluoride, 5-Chloro-alpha,alpha,alpha-trifluoro-m-tolunitrile
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-chloro-5-trifluoromethylbenzonitrile

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/3-chloro-5-trifluoromethylbenzonitrile
pictograms GHS06
hstatements H311;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart