APO455837749 Apollo Scientific Ltd

4-Bromo-2-(trifluoromethoxy)benzonitrile

  • MDL number MFCD12761591
  • Cas Number : 1187983-97-6
  • Molecular Weight : 266.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-bromo-2-trifluoromethoxybenzonitrile

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-2-trifluoromethoxybenzonitrile
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart