APO455837750 Apollo Scientific Ltd

5-Bromo-2-(trifluoromethoxy)benzonitrile

  • MDL number MFCD13185875
  • Cas Number : 1210906-15-2
  • Molecular Weight : 266.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Bromo-2-cyano-1-(trifluoromethoxy)benzene
Purity 99
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-bromo-2-trifluoromethoxybenzonitrile

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-2-trifluoromethoxybenzonitrile
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart