APO455837751 Apollo Scientific Ltd

2,5-Difluoro-4-nitrophenol

  • MDL number MFCD04039297
  • Cas Number : 120103-18-6
  • Molecular Weight : 175.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,5-difluoro-4-nitrophenol
Inchi Code 1S/C6H3F2NO3/c7-3-2-6(10)4(8)1-5(3)9(11)12/h1-2,10H
Inchi Key WOQWWNJQZLYLMC-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/25-difluoro-4-nitrophenol

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/25-difluoro-4-nitrophenol
pictograms GHS05;GHS07
hstatements H302;H315;H318;H335
Availability details for
Quantity
  • Availability
  • Add to Cart