APO455837752 Apollo Scientific Ltd

Methyl 4-fluoro-2-methyl-5-nitrobenzoate

  • MDL number MFCD14581697
  • Cas Number : 1163287-01-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 68-71
Synonym 5-Fluoro-2-(methoxycarbonyl)-4-nitrotoluene, 2-Fluoro-5-(methoxycarbonyl)-4-methylnitrobenzene
Purity 98
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=46810

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=46810
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart