APO455837755 Apollo Scientific Ltd

2,3,5,6-Tetrafluorotoluene

  • MDL number MFCD00000313
  • Cas Number : 5230-78-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1,2,4,5-Tetrafluoro-3-methylbenzene
Purity 98
Density 1.347
Flash Point 25
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 125-126
Product Link http://www.apolloscientific.co.uk/display_item.php?id=4881

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=4881
pictograms GHS02;GHS07
hstatements H226;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart