APO455837757 Apollo Scientific Ltd

1H,2H-Heptafluoro(1-iodo-3-methylbut-1-ene)

  • MDL number MFCD00155985
  • Cas Number : 243139-61-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-Iodo-3,4,4,4-tetrafluoro-3-(trifluoromethyl)but-1-ene, 3,4,4,4-Tetrafluoro-1-iodo-3-(trifluoromethyl)-1-butene
Purity 97
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 98
Product Link http://www.apolloscientific.co.uk/display_item.php?id=4895

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=4895
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart