APO455837759 Apollo Scientific Ltd

4,5,5,5-Tetrafluoro-4-(trifluoromethyl)pent-2-enoic acid

  • MDL number MFCD09998187
  • Cas Number : 243139-64-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-(Perfluoroisopropyl)acrylic acid, 3-(Heptafluoroprop-2-yl)prop-2-enoic acid
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=4899

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=4899
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart