APO455837762 Apollo Scientific Ltd

4-Bromo-2-fluorophenacyl bromide

  • MDL number MFCD11847024
  • Cas Number : 869569-77-7
  • Molecular Weight : 295.93

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 79-80
Synonym 2,4'-Dibromo-2'-fluoroacetophenone, 2-Bromo-1-(4-bromo-2-fluorophenyl)ethan-1-one
Purity 98
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-bromo-2-fluorophenacyl-bromide

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-2-fluorophenacyl-bromide
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart