APO455837766 Apollo Scientific Ltd

2,6-Dichloro-5-fluoronicotinamide

  • MDL number MFCD01571365
  • Cas Number : 113237-20-0
  • Molecular Weight : 209.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 161-164
Synonym 2,6-Dichloro-5-fluoropyridine-3-carboxamide, 3-Carbamoyl-2,6-dichloro-5-fluoropyridine
Purity 97
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/26-dichloro-5-fluoronicotinamide

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/26-dichloro-5-fluoronicotinamide
pictograms GHS07
hstatements H302+H312+H332;H319
Availability details for
Quantity
  • Availability
  • Add to Cart