APO455837770 Apollo Scientific Ltd

4-Amino-2-fluoro-5-nitrobenzotrifluoride

  • MDL number MFCD09907681
  • Cas Number : 428871-73-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 137-140
Synonym 5-Fluoro-2-nitro-4-(trifluoromethyl)aniline, 2-Amino-4-fluoro-5-(trifluoromethyl)nitrobenzene
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=47019

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=47019
pictograms GHS06
hstatements H301+H311+H331
Availability details for
Quantity
  • Availability
  • Add to Cart