APO455837771 Apollo Scientific Ltd

3-Acetyl-2-fluoropyridine

  • MDL number MFCD07375072
  • Cas Number : 79574-70-2
  • Molecular Weight : 139.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-(2-Fluoropyridin-3-yl)ethan-1-one, 3-Ethanoyl-2-fluoropyridine
Purity 97
Density 1.18
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 85/10mm
Product Link https://store.apolloscientific.co.uk/product/3-acetyl-2-fluoropyridine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-acetyl-2-fluoropyridine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart