APO455837775 Apollo Scientific Ltd

3-(Trifluoromethoxy)toluene

  • MDL number MFCD07366526
  • Cas Number : 705-44-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=9252

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=9252
pictograms GHS02;GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart