APO455837781 Apollo Scientific Ltd

3,4,5-Trichlorobenzotrifluoride

  • MDL number MFCD00035985
  • Cas Number : 50594-82-6
  • Molecular Weight : 249.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point -10 to -8
Synonym 1,2,3-Trichloro-5-(trifluoromethyl)benzene, 3,4,5-Trichloro-alpha,alpha,alpha-trifluorotoluene
Purity 98
Density 1.60
Flash Point 98
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 200-206
Product Link https://store.apolloscientific.co.uk/product/345-trichlorobenzotrifluoride

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/345-trichlorobenzotrifluoride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart