APO455837789 Apollo Scientific Ltd

3-(Ethoxycarbonyl)-2-fluorobenzeneboronic acid

  • MDL number MFCD08064050
  • Cas Number : 1072952-52-3
  • Molecular Weight : 211.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Ethyl 3-borono-2-fluorobenzoate
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-ethoxycarbonyl-2-fluorobenzeneboronic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-ethoxycarbonyl-2-fluorobenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart