APO455837791 Apollo Scientific Ltd

6-Fluoronicotinaldehyde

  • MDL number MFCD08064049
  • Cas Number : 677728-92-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 6-Fluoro-3-formylpyridine, 6-Fluoropyridine-3-carboxaldehyde, 2-Fluoro-5-formylpyridine
Purity 97
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/6-fluoronicotinaldehyde

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/6-fluoronicotinaldehyde
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart