APO455837794 Apollo Scientific Ltd

5-Fluoropyridine-2-carboxaldehyde

  • MDL number MFCD07781242
  • Cas Number : 31181-88-1
  • Molecular Weight : 125.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 31-33
Iupac Name 5-fluoro-2-pyridinecarbaldehyde
Inchi Code 1S/C6H4FNO/c7-5-1-2-6(4-9)8-3-5/h1-4H
Inchi Key IACCXWQKIQUVFQ-UHFFFAOYSA-N

Description

Synonym 5-Fluoro-2-formylpyridine, 5-Fluoropicolinaldehyde
Flash Point 72-74/21mm
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 31-33
Product Link https://store.apolloscientific.co.uk/product/5-fluoropyridine-2-carboxaldehyde

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-fluoropyridine-2-carboxaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart