APO455837795 Apollo Scientific Ltd

3-Fluoro-2-hydroxybenzeneboronic acid

  • MDL number MFCD07781238
  • Cas Number : 259209-24-0
  • Molecular Weight : 155.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Borono-6-fluorophenol
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-fluoro-2-hydroxybenzeneboronic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-fluoro-2-hydroxybenzeneboronic-acid
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart