APO455837799 Apollo Scientific Ltd

6-Fluoronicotinoyl chloride

  • MDL number MFCD07781233
  • Cas Number : 65352-94-5
  • Molecular Weight : 159.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 275-278
Iupac Name 6-fluoronicotinoyl chloride
Inchi Code 1S/C6H3ClFNO/c7-6(10)4-1-2-5(8)9-3-4/h1-3H
Inchi Key IPOCOANFUVSCLZ-UHFFFAOYSA-N

Description

Synonym 2-Fluoropyridine-5-carbonyl chloride, 6-Fluoropyridine-3-carbonyl chloride, 5-(Chlorocarbonyl)-2-fluoropyridine
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 275-278
Product Link https://store.apolloscientific.co.uk/product/6-fluoronicotinoyl-chloride

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/6-fluoronicotinoyl-chloride
pictograms GHS05;GHS07
hstatements H302;H314
Availability details for
Quantity
  • Availability
  • Add to Cart