APO455837800 Apollo Scientific Ltd

3-Fluoropyridine-2-carboxylic acid

  • MDL number MFCD04114108
  • Cas Number : 152126-31-3
  • Molecular Weight : 141.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 167-169(dec.)
Iupac Name 3-fluoro-2-pyridinecarboxylic acid
Inchi Code 1S/C6H4FNO2/c7-4-2-1-3-8-5(4)6(9)10/h1-3H,(H,9,10)
Inchi Key IRERRSXDWUCFIY-UHFFFAOYSA-N

Description

Synonym 3-Fluoropicolinic acid, 2-Carboxy-3-fluoropyridine
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 167-169(dec.)
Product Link https://store.apolloscientific.co.uk/product/3-fluoropyridine-2-carboxylic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-fluoropyridine-2-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart