APO455837802 Apollo Scientific Ltd

2-Bromo-4-fluorostyrene

  • MDL number MFCD14892603
  • Cas Number : 1221684-51-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=47011

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=47011
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart