APO455837806 Apollo Scientific Ltd

4,5,7-Trichloro-2-(trifluoromethyl)quinoline

  • MDL number MFCD00153279
  • Cas Number : 175203-41-5
  • Molecular Weight : 300.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 111-114
Synonym 4,5,7-Trichloro-2-(trifluoromethyl)-1-azanaphthalene
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/457-trichloro-2-trifluoromethylquinoline

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/457-trichloro-2-trifluoromethylquinoline
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart