APO455837811 Apollo Scientific Ltd

2-(Aminomethyl)-3-chloro-5-(trifluoromethyl)pyridine hydrochloride

  • MDL number MFCD00210134
  • Cas Number : 326476-49-7
  • Molecular Weight : 247.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 226-229
Synonym [3-Chloro-5-(trifluoromethyl)pyridin-2-yl]methylamine hydrochloride, 3-Chloro-5-(trifluoromethyl)picolinylamine hydrochloride
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-aminomethyl-3-chloro-5-trifluoromethylpyridine-hydrochloride

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-aminomethyl-3-chloro-5-trifluoromethylpyridine-hydrochloride
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart