APO455837812 Apollo Scientific Ltd

(3-Fluorophenyl)methanesulphonyl chloride

  • MDL number MFCD04117456
  • Cas Number : 24974-72-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 39-41
Purity 97
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/3-fluorophenylmethanesulphonyl-chloride-97

Safety Information

signalword Danger
pcodes P280;P271;P260
MSDS https://store.apolloscientific.co.uk/product/3-fluorophenylmethanesulphonyl-chloride-97
pictograms GHS05
hstatements H314;EUH014;EUH029
Availability details for
Quantity
  • Availability
  • Add to Cart