APO455837814 Apollo Scientific Ltd

5-Fluoronicotinonitrile

  • MDL number MFCD09027295
  • Cas Number : 696-42-4
  • Molecular Weight : 122.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 50-54
Synonym 3-Cyano-5-fluoropyridine, 5-Fluoropyridine-3-carbonitrile
Purity 97
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-fluoronicotinonitrile

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-fluoronicotinonitrile
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart