APO455837816 Apollo Scientific Ltd

5-Bromo-2-fluorophenylhydrazine hydrochloride

  • MDL number MFCD03094411
  • Cas Number : 214916-08-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Bromo-1-fluoro-2-hydrazinobenzene hydrochloride
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/5-bromo-2-fluorophenylhydrazine-hydrochloride

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-2-fluorophenylhydrazine-hydrochloride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart