APO455837824 Apollo Scientific Ltd

2-Fluoro-4-(methylsulphonyl)benzeneboronic acid

  • MDL number MFCD09027248
  • Cas Number : 957060-85-4
  • Molecular Weight : 218.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 162-168
Purity 98
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-fluoro-4-methylsulphonylbenzeneboronic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-fluoro-4-methylsulphonylbenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart