APO455837826 Apollo Scientific Ltd

2-Oxo-5-(trifluoromethoxy)indoline

  • MDL number MFCD09027275
  • Cas Number : 403853-48-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-(Trifluoromethoxy)-1,3-dihydro-2H-indol-2-one
Purity 98
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=36389

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=36389
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart