APO455837830 Apollo Scientific Ltd

2'-Fluoro-4'-iodoacetanilide

  • MDL number MFCD04039298
  • Cas Number : 97760-94-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N-(2-Fluoro-4-iodophenyl)acetamide, N-(2-Fluoro-4-iodophenyl)ethanamide
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=8185

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=8185
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart