APO455837831 Apollo Scientific Ltd

Trifluoroacetaldehyde methyl hemiacetal

  • MDL number MFCD00013572
  • Cas Number : 431-46-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-Methoxy-2,2,2-trifluoroethan-1-ol, 2-Hydroxy-2-methoxy-1,1,1-trifluoroethane
Density 1.26
Flash Point 42
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 96
Product Link http://www.apolloscientific.co.uk/display_item.php?id=4957

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=4957
pictograms GHS02;GHS07
hstatements H226;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart